Nitrite test strips, Dosatest
Lieferant: VWR Chemicals
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
85705.601EA 33 EUR InStock
85705.601
Nitrite test strips, Dosatest
Umwelt-Prüfkits
Semi-quantitative test strips for the determination of nitrite.
  • Ideal for rapid and easy analysis of nitrite directly on-site
  • No maintenance or calibration is required
Colour change: Yellow to red
Lieferumfang: Delivered as 100 test strips in a tube, instruction leaflet in a box
Achtung! Bitte beachten: Strongly acidic solutions must be diluted with dilute sodium hydroxide and strongly alkaline samples with dilute sulphuric acid to pH 4 to 6.
Order Now
PRODUKT
TECHNISCHE DATEN
WEITERE INFORMATIONEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge