Total Iron Test Strips, Dosatest, VWR®
Lieferant: VWR Chemicals
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
85709.601EA 28.1 EUR InStock
85709.601 85713.601
Total Iron Test Strips, Dosatest, VWR®
Umwelt-Prüfkits
Semi-quantitative test strips for the determination of iron.
  • Ideal for rapid and easy analysis of iron directly on-site
  • No maintenance or calibration is required
Colour change 0 - 1000: White to dark red.
Colour change 0 - 100: White to blue violet
Lieferumfang: Delivered as 100 test strips in a tube with instruction leaflet.
Achtung! Bitte beachten: For the lower concentration test strip, the coloration produced by some heavy metals in higher concentrations can be eliminated by adding cadmium powder.
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge