VWR® ARISTAR®, Ion chromatography anion multi-element standard solution, Certified Reference Material
Lieferant: VWR Chemicals
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
85783.154EA 163 EUR InStock
85783.154
VWR® ARISTAR®, Ion chromatography anion multi-element standard solution, Certified Reference Material
Standards Anionische Standards Anionische Multi-Element-Standards
This certified reference material is produced using a high purity starting material, acid from sub-boiling and 18 MOhm deionized water and filtered through a 0.2 micron filter.
10 mg/kg each of: F⁻, Cl⁻, Br-, NO₃⁻, PO₄³⁻, SO₄²⁻ in H2O
Zertifizierungen: CRM ISO 17034 and ISO 17025. Traceable to NIST.
Order Now
PRODUKT
TECHNISCHE DATEN
WEITERE INFORMATIONEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge