Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Azur A 70% (Farbstoffgehalt) certified by the Biological Stain Commission, Sigma-Aldrich®

Lieferant: SIGMA ALDRICH MICROSCOPY

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

Achtung

M-Clarity™ Qualitätsstufe = MQ200

861049-25G 861049-5G
861049-25GEA 188 EUR InStock
861049-25G 861049-5G
Azur A 70% (Farbstoffgehalt) certified by the Biological Stain Commission, Sigma-Aldrich®
Azur A
The combination of azure A and silver staining is used for the quantification of glycosaminoglycans in agarose gels. It is also used for the staining of chromosomes in the nucleus. Azure A is a main component of the azure B/polychromed methylene blue-eosin stains family, also referred to as the Romanowsky stain. Azure A is a basic cationic dye.

Azure A has been used to determine the amount of charged dextran sulfate in the nanoparticles for the study involving incorporation of heparin-binding proteins into preformed dextran sulfate-chitosan nanoparticles. Certified for use in Lillie′s modified Nocht′s method and the NCCLS method for blood smears.


Formel: C₁₄H₁₄ClN₃S
MW: 291.797 g/mol
Melting Pt: 290 °C (Zers., Lit.)
Storage Temperature: Room Temperature
MDL Number: MFCD00012112
CAS-Nummer: 531-53-3
EINECS: 208-510-7
https://qa.digitalassets.avantorsciences.com/adaptivemedia/rendition?id=e2e0283a1a435df57e451656fa78c07fe5f5402b&vid=e2e0283a1a435df57e451656fa78c07fe5f5402b&prid=web&clid=SAPDAM
Order Now
Spezifikation Testergebnisse
Appearance (Colour) Blue to Very Dark Blue to Very Dark Green
Appearance (Form) Powder or Crystals
Infrared spectrum Conforms to Structure
Wave Length, c = 0.003g/L; Water 620 - 634 nm
Extinction Coefficient ≥11000
Wave Length 242 - 246 nm
Solubility (Turbidity), c = 1 mg/ml; Water Turbid
Solubility (Colour) Dark Blue to Very Dark Blue and Dark Purple to Very Dark Purple and Blue-Purple
Biological Stain Commision, Commission Certified Number: Pass
Dye Content Approximately 70%

 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge