Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Magnesiumchlorid Hexahydrat 98,0-101,0% USP, Ph. Eur., BP

Lieferant: VWR Chemicals

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:


87060.290EA 51.5 EUR InStock
87060.290 87060.360 87060.460
Magnesiumchlorid Hexahydrat 98,0-101,0% USP, Ph. Eur., BP
Magnesiumchlorid Hexahydrat
Achtung! Bitte beachten: Not for use as Active Pharmaceutical Ingredient (API)


Formel: MgCl₂·6H₂O
Siedepunkt: 1412 °C (1013 hPa)
Melting Pt: 117 °C
Dichte: 1,57 g/cm³ (25 °C)
Storage Temperature: Raumtemperatur
MDL Number: MFCD00149781
CAS-Nummer: 7791-18-6
EINECS: 232-094-6
Merck Index: 13,05684
Order Now
Spezifikation Testergebnisse
Appearance Colourless crystals
Heavy metals (as Pb) ≤ 10 ppm
Residual solvents Unlikely by manuf.process
Residues of metal catalysts or reagents Unlikely by manuf.process
As (Arsenic) ≤ 2 ppm
Cd (Cadmium) ≤ 1 ppm
Co (Cobalt) ≤ 1 ppm
Hg (Mercury) ≤ 2 ppm
Ni (Nickel) ≤ 6 ppm
Pb (Lead) ≤ 2 ppm
V (Vanadium) ≤ 10 ppm
Conforms to Ph.Eur./USP/BP Passes test
Ph.Eur./BP specifications: .
Assay [Ph.Eur./BP] 98.0 - 101.0 %
Identification A Passes test USP
Identification B Passes test USP
Identification C Passes test Ph.Eur./BP
Appearance of solution Passes test Ph.Eur./BP
Acidity or alkalinity Passes test Ph.Eur./BP
Br (Bromide) [Ph.Eur./BP] ≤ 500 ppm
SO4 (Sulphate) [Ph.Eur./BP] ≤ 100 ppm
Ca (Calcium) [Ph.Eur./BP] ≤ 0.1 %
Fe (Iron) [Ph.Eur./BP] ≤ 10 ppm
Water [Ph.Eur./BP] 51.0 - 55.0 %
USP specifications: .
Assay [USP] 98.0 - 101.0 %
Insoluble matter [USP] ≤ 0.005 %
SO4 (Sulphate) [USP] ≤ 0.005 %
Ba (Barium) Passes test USP
Limit of calcium [USP] ≤ 0.01 %
K (Potassium) Passes test USP
pH (25°C; 50 mg/ml) [USP] 4.5 - 7.0

 

PRODUKT

TECHNISCHE DATEN

WEITERE INFORMATIONEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge