BRP Indicator solution Reag. Ph. Eur. 1013000
Lieferant: VWR Chemicals
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
87799.180EA 52.5 EUR InStock
87799.180
BRP Indicator solution Reag. Ph. Eur. 1013000
BRP Indicator solution
Solution of Bromothymol Blue R, Methyl Red R and Phenolphtthalein R in alcohol
Order Now
PRODUKT
TECHNISCHE DATEN
WEITERE INFORMATIONEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge