VWR®, Residual Solvents Class 1 According to Ph. Eur. Versions 8.8 and 10
Lieferant: VWR Chemicals
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
88215.001EA 145 EUR InStock
88215.001
VWR®, Residual Solvents Class 1 According to Ph. Eur. Versions 8.8 and 10
Standards Organische Standards
The level of volatile organic solvents in pharmaceutical raw materials or finished product is defined and monitored. Residual solvents are classified according to their risk assessment classes. A selection of residual solvent standards is now available from Avantor.
Residual Solvents Class 1 acc to Ph Eur Ver 8.8: 5 components; benzene 4 mg/ml; 1,2-dichloroethane 10 mg/ml ; tetrachloromethane 8 mg/ml; 1,1-dichloroethene 16 mg/ml; 1,1,1-trichloroethane 20 mg/ml in imethylsulphoxide
Residual Solvents Class 1 acc. to Ph Eur Ver 10: 5 components; benzene 2 mg/L; tetrachloromethane (carbon tetrachloride) 4 mg/L; 1,2-dichloroethane 5 mg/L; 1,1-dichloroethene 8 mg/L; 1,1,1-trichloroethane 1500 mg/L in dimethyl sulphoxide.
Zertifizierungen: All Solutions are Certified Reference Materials, produced and calibrated under a quality system that is:
- ISO 9001 certified.
- accredited according to ISO/IEC 17025 - Testing
- accredited according to ISO 17034 - Reference Material Producer
Order Now
PRODUKT
TECHNISCHE DATEN
WEITERE INFORMATIONEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge