88285.14
88285.22
88285.14EA
176
EUR
InStock
88285.14
88285.22
Blei(II)tellurid ≥99,999% (Metall-Basis)
Blei(II)tellurid
Purity: 99.999%
CAS No.: 1314-91-6
Molecular Formula: PbTe
Formula Weight: 334.79
Physical Form: 15mm & Down Pieces
Density: 8.164
Melting Point: 917°
Merck Reference: 14,5422
UN No.: 3284
MDL No.: MFCD00016274