88777.14
88777.22
88777.14EA
162
EUR
InStock
88777.14
88777.22
Siliciumdioxid ≥99,995% (Metall-Basis)
Siliciumdioxid
Purity: 99.995%
CAS No.: 7631-86-9
Molecular Formula: SiO2
Formula Weight: 60.09
Physical Form: -40 Mesh Powder
Density: 2.2
Appearance: White
Odour: Odourless
Melting Point: 1710°
Boiling Point: 2230°
Merck Reference: 14,8493
MDL No.: MFCD00011232