88960.03
88960.06
88960.03EA
176
EUR
InStock
88960.03
88960.06
Tetraammineplatinum(II) nitrate
Tetraammineplatinum(II) nitrate
CAS No.: 20634-12-2
Molecular Formula: H12N6O6Pt
Formula Weight: 387.21
Physical Form: Powder/chunks
Appearance: White
Melting Point: 262° dec.
UN No.: UN1477
MDL No.: MFCD00011624