89020.30
89020.18
89020.A1
89020.30EA
200
EUR
InStock
89020.30
89020.18
89020.A1
Nickel(II)sulfamat Hydrat
Nickel(II)sulfamat Hydrat
Nickel(II) sulfamate is used as an electro plating and surface treating agent. It finds application in metal coloring and casting.
CAS No.: 13770-89-3
Molecular Formula: Ni(SO3NH2)2.xH2O
Formula Weight: 250.86(anhy)
Physical Form: Crystalline
Appearance: Blue
Odour: Odourless
UN No.: 3077
MDL No.: MFCD00137261