VWR®, Native PFAS Reference Standard, CRM
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
89086.010EA 0 EUR InStock
89086.010
VWR®, Native PFAS Reference Standard, CRM
Standards Umweltanalyse-Standards
This CRM contains the 24 PFAS based on the newest publication of USEPA Method 8327 which is suitable for testing PFAS in surface water, ground water and wastewater matrices. The two CRMs (89086 and 89087) are offered at a high and a low concentration to meet the specific needs of your testing.
Contains:
Perfluorobutane-1-sulfonic acid (CAS 375-73-5)
Perfluoropentanesulfonic acid (CAS 2706-91-4
Perfluorohexane-1-sulfonic acid (Linear and Branched) (CAS 355-46-4
Perfluoroheptanesulfonic acid (CAS 375-92-8)
Perfluorooctane-1-sulfonic acid (Linear and branched) (CAS 1763-23-1)
Perfluorononanesulfonic acid (CAS 68259-12-1)
Perfluorodecane-1-sulfonic acid (CAS 335-77-3)
1H,1H,2H,2H-Perfluorohexanesulfonic acid (CAS 757124-72-4)
1H,1H,2H,2H-Perfluorooctane sulfonic acid (CAS 27619-97-2)
1H,1H,2H,2H-Perfluorodecanesulfonic acid (CAS 39108-34-4)
Perfluoro-n-butanoic acid (CAS 375-22-4)
Perfluoro-n-pentanoic acid (CAS 2706-90-3)
Perfluoro-n-hexanoic acid (CAS 307-24-4)
Perfluoro-n-heptanoic acid (CAS 375-85-9)
Perfluoro-n-octanoic acid (CAS 335-67-1
Perfluoro-n-nonanoic acid (CAS 375-95-1)
Perfluoro-n-decanoic acid (CAS 335-76-2)
Perfluoro-n-undecanoic acid (CAS 2058-94-8)
Perfluoro-n-dodecanoic acid (CAS 307-55-1)
Perfluoro-n-tridecanoic acid (CAS 72629-94-8)
Perfluoro-n-tetradecanoic acid (CAS 376-06-7)
N-ethylperfluoro-1-octanesulfonamidoacetic acid (CAS 2991-50-6)
N-methylperfluoro-1-octanesulfonamidoacetic acid (CAS 2355-31-9)
Perfluorooctane sulfonamide (CAS 754-91-6)
Zertifizierungen: ISO 17034
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge