89153.14
89153.14EA
25.7
EUR
InStock
89153.14
Trimethoxyboroxin
2,4,6-Trimethoxyboroxin
CAS No.: 102-24-9
Molecular Formula: (CH3O)3B3O3
Formula Weight: 173.53
Flash Point: 14°(57°F)
Physical Form: Liquid
Density: 1.216
Refractive Index: 1.3996
Sensitivity: Air and Moisture Sensitive
Odour: Strong
Melting Point: 10-11°
Boiling Point: 130°
Beilstein Reference: 1709158
UN No.: UN2924
MDL No.: MFCD00005946