Phytochemical standard, Ginkgotoxin, analytical standard, Supelco®
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
89960-10MG
89960-10MGEA 67.7 EUR InStock
89960-10MG
Phytochemical standard, Ginkgotoxin, analytical standard, Supelco®
Standards Organische Standards
Ginkgotoxin is an alkaloid, which occurs in the seeds and leaves of Ginkgo biloba. It is also a potential antivitamin, structurally related to vitamin B6.
Ginkgotoxin may be used as a standard in the determination of ginkgotoxin in selected chinese herbs and plants using high-performance liquid chromatography (HPLC) or gas chromatography coupled with mass spectrometry (GC-MS).
https://qa.digitalassets.avantorsciences.com/adaptivemedia/rendition?id=03c4c177d77bf6c5bd1f963d5cf7df1312eae009&vid=03c4c177d77bf6c5bd1f963d5cf7df1312eae009&prid=web&clid=SAPDAM
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge