Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

8-Aminonaphthalen-1,3,6-trisulfonsäure Dinatriumsalz fluoreszierender Farbstoff

Lieferant: BIOTIUM

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:


90010
90010.EA 118 EUR InStock
90010.
8-Aminonaphthalen-1,3,6-trisulfonsäure Dinatriumsalz fluoreszierender Farbstoff
8-Aminonaphthalene-1,3,6-trisulphonic acid disodium salt
ANTS (8-aminonaphthalene-1,3,6-trisulfonic acid, disodium salt) is a highly negatively charged dye with an amino group that can be coupled to an aldehyde or ketone group to form an unstable Schiff base. The Schiff base is usually chemically reduced by sodium borohydride (NaBH4) or sodium cyanoborohydride (NaB(CN)H3) to form a stable linkage. This labeling technique has been widely used for the labeling and subsequent sequencing of oligosaccharides and glycoproteins. The negative charges of the dye facilitate the electrophoretic separation of the degradation products of carbohydrate polymers.

  • λEx/λEm = 353/520 nm
  • ε= 65,000
  • Light yellow solid soluble in water and DMSO
  • Store at 4°C and protect from light, especially in solution

ANTS has also been used together with the fluorescent quencher DPX (80012) for the studies of membrane fusion or permeability. The mixture of ANTS-DPX is minimally fluorescent initially, but becomes increasingly more fluorescent upon dilution (i.e., membrane fusion or leakage).


Formel: C₁₀H₉NO₉S₃
Storage Temperature: Kühlschrank
MDL Number: MFCD00070572
CAS-Nummer: 5398-34-5
https://qa.digitalassets.avantorsciences.com/adaptivemedia/rendition?id=6600f03eeb2b58ccbcd6ff9031d11c12400513b2&vid=6600f03eeb2b58ccbcd6ff9031d11c12400513b2&prid=web&clid=SAPDAM
Order Now

 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge