Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

8-Aminopyrene-1,3,6-trisulphonic acid trisodium salt fluoreszierender Farbstoff

Lieferant: BIOTIUM

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

Achtung

90028
90028.EA 232 EUR InStock
90028.
8-Aminopyrene-1,3,6-trisulphonic acid trisodium salt fluoreszierender Farbstoff
8-Aminopyrene-1,3,6-trisulphonic acid trisodium salt
APTS is a very useful green fluorescent dye for labeling glycoproteins or sugar molecules in general. The labeling occurs via reductive amination which involves the formation of a Schiff base between the amine of APTS and the aldehyde or ketone of the sugar, followed by the reduction of the Schiff base linkage to a stable carbon-nitrogen bond. The multi-anionic nature of the dye makes it ideal for the studies of carbohydrate molecules by high resolution capillary electrophoresis.

  • λ Ex/λ Em: 424/505 nm (before conjugation)
  • Yellow solid soluble in water
  • Store at 4°C and protect from light


Formel: C₁₆H₈NNa₃O₉S₃
Storage Temperature: Kühlschrank
MDL Number: MFCD00674494
CAS-Nummer: 196504-57-1
https://qa.digitalassets.avantorsciences.com/adaptivemedia/rendition?id=09884deb23ea1e292890e9582671db25ac98fe62&vid=09884deb23ea1e292890e9582671db25ac98fe62&prid=web&clid=SAPDAM
Order Now

 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge