90079
90063
90059
90050
90078
90067
90068
90069
90080
90061
90053
90051
90058
90054
90052
90064
90064-1
90065
90065-1
90066
90066-1
92010
92011
92012
90083
90111
90112
90113
92167
92168
92169
92176
90079.EA
298
EUR
InStock
90079.
90063.
90059.
90050.
90078.
90067.
90068.
90069.
90080.
90061.
90053.
90051.
90058.
90054.
90052.
90064.
90064-1
90065.
90065-1
90066.
90066-1
92010.
92011.
92012.
90083.
90111.
90112.
90113.
92167.
92168.
92169.
92176.
Reactive biotin compounds
Proteinkonjugate und Markierungen
Biotium offers reactive biotin compounds with a variety of reactive functional groups.
- Reagents for biotinylation of proteins and other biomolecules
- Full selection of reactive functional groups
- Variety of linker lengths available for improved water solubility and streptavidin binding
Amine reactive:
Succinimidyl esters react with primary and secondary amines and can be used to label proteins or peptides
Thiol reactive:
Maleimide and iodoacetamide react with thiol groups
MTSEA selectively labels thiols under mild conditions
TS biotin compounds allow selective labeling of cell surface thiols
Other reactive chemistries:
Amine groups can be reacted with activated carboxylic acids
Cadaverine can be coupled to carboxylic acids, DNA, and other biomolecules
Cadaverine free base can be directly coupled to activated carboxylic acids without the use of additional base
Hydrazides react with aldehydes or ketones to form unstable hydrazone linkages
Aminooxy reacts with aldehydes or ketones to form stable oxime bonds, and may be preferable to hydrazides for labeling
Tyramides can be used for HRP-mediated tyramide signal amplification (TSA) reactions
Alkynes react with azides by copper(I)-catalyzed 1,3-dipolar Huisgen cycloaddition
BCN groups react with azides by copper-free 1,3-dipolar Huisgen cycloaddition
Non-reactive free acid forms:
Biotin-X free acid (6-((Biotinoyl)amino)hexanoic acid) can be easily converted to the mixed anhydride form, which is more reactive than the succinimidyl ester and thus can be used to react with aromatic amines or sterically hindered amines.
Biotin-XX free acid (6-((6-((Biotinoyl)amino)hexanoyl)amino)hexanoic acid) can be easily converted to the mixed anhydride form, which is more reactive than the succinimidyl ester and thus can be used to react with aromatic amines or sterically hindered amines.
Note: A full range of fluorescent CF™ Dyes are also available.