Organic reference standard, Fenofibric acid, analytical standard, Supelco®
Lieferant: SUPELCO
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
90568-10MG
90568-10MGEA 91.6 EUR InStock
90568-10MG
Organic reference standard, Fenofibric acid, analytical standard, Supelco®
Standards Organische Standards
Used as a reference standard in determination of fenofibric acid in samples of human plasma using high performance liquid chromatography (HPLC) and liquid chromatography coupled with tandem mass spectrometry (LC-MS-MS).
https://qa.digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ce64e9c732507d863c7f425cdbc3aa408ad045b7&vid=ce64e9c732507d863c7f425cdbc3aa408ad045b7&prid=web&clid=SAPDAM
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge