Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Kaliumbromid ≥99,9995% (Metall-Basis), TraceSELECT™, Fluka™

Lieferant: HONEYWELL

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:


90737-100G
90737-100GEA 259.32 EUR InStock
90737-100G
Kaliumbromid ≥99,9995% (Metall-Basis), TraceSELECT™, Fluka™
Kaliumbromid


Formel: KBr
Siedepunkt: 1,383 °C (1.013 hPa)
Melting Pt: 734 °C
Dichte: 2,73 g/cm3 (20 °C)
Storage Temperature: Raumtemperatur
MDL Number: MFCD00011358
CAS-Nummer: 7758-02-3
Order Now
Spezifikation Testergebnisse
Aluminium (Al) Max. 0.02 ppm
Arsenic (As) Max. 0.01 ppm
Barium (Ba) Max. 0.5 ppm
Calcium (Ca) Max. 0.1 ppm
Cadmium (Cd) Max. 0.005 ppm
Cerium (Ce) Max. 0.005 ppm
Cobalt (Co) Max. 0.005 ppm
Chromium (Cr) Max. 0.005 ppm
Copper (Cu) Max. 0.005 ppm
Iron (Fe) Max. 0.01 ppm
Mercury (Hg) Max. 5 ppb
Iridium (Ir) Max. 0.5 ppm
Lanthanum (La) Max. 0.005 ppm
Lithium (Li) Max. 0.5 ppm
Magnesium (Mg) Max. 0.05 ppm
Manganese (Mn) Max. 0.005 ppm
Sodium (Na) Max. 5 ppm
Nickel (Ni) Max. 0.005 ppm
Lead (Pb) Max. 0.005 ppm
Palladium (Pd) Max. 0.5 ppm
Platinum (Pt) Max. 0.5 ppm
Rhodium (Rh) Max. 0.5 ppm
Ruthenium (Ru) Max. 0.5 ppm
Antimony (Sb) Max. 0.01 ppm
Scandium (Sc) Max. 0.005 ppm
Selenium (Se) Max. 0.01 ppm
Samarium (Sm) Max. 0.005 ppm
Strontium (Sr) Max. 0.05 ppm
Thallium (Tl) Max. 0.01 ppm
Vanadium (V) Max. 0.5 ppm
Yttrium (Y) Max. 0.005 ppm
Zinc (Zn) Max. 0.01 ppm
Bromate (BrO₃) Max. 10 ppm
Chloride (Cl) Max. 500 ppm
Iodide (I) Max. 200 ppm
Sulfate (SO₄) Max. 10 ppm
Total N Max. 2 ppm

 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge