VWR® PESTINORM®, Gas Chromatography (GC) Certified Reference Material
Lieferant: VWR Chemicals
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
91000.050EA 215 EUR InStock
91000.050 91001.050 91002.050 91003.050 91004.050 91005.050 91006.050 91007.050 91008.050 91009.050 91010.050 91012.050 91013.050 91014.050 91015.050 91016.050 91017.050 91018.050 91019.050 91020.010 91022.001 91023.050 91030.050 91032.050 91033.050 91034.050 91035.050 91036.050 91037.050 91038.050 91039.050 91040.050 91041.050 91042.050 91043.050 91044.050 91045.050 91046.050 91047.050 91048.050 91049.050 91050.050 91051.050 91052.050 91053.050 91054.050 91055.050 91056.050 91057.050 91058.050 91059.050 91060.050 91061.050 91062.050 91063.050 91064.050 91065.050 91066.050 91067.050 91068.050 91069.050 91070.050 91071.050
VWR® PESTINORM®, Gas Chromatography (GC) Certified Reference Material
Standards Gaschromatographie-Standards
Due to its excellent separation capacity and its high sensitivity gas chromatography is a widely used technique for analytical purposes. In order to get proper identification and quantification results of the analysed compounds, reference standards are needed. These PESTINORM® reference substances can be used for the characterisation of GC column properties. In addition they may also be used for the identification of unknown compounds in a gas chromatogram or as standards in quantitative GC analysis.
  • All products are CRM acc ISO/IEC 17025 and ISO 17034
  • All products supplied with a Certificate of Analysis and batch-specific chromatogram
  • Assay typically min 99,0% in many cases over 99,5%
  • 24 month shelf life
Intended uses:
- For calibration of TLC, GC-FID, GC-TCD, GC-ECD, GC-MS, GC-MS-MS, LC-UV, LC-MS and LC-MS-MS
- Validation of analytical methods
- Preparation of 'working reference samples'
- Detection limit and linearity studies
Order Now
PRODUKT
TECHNISCHE DATEN
WEITERE INFORMATIONEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge