91000.050EA
215
EUR
InStock
91000.050
91001.050
91002.050
91003.050
91004.050
91005.050
91006.050
91007.050
91008.050
91009.050
91010.050
91012.050
91013.050
91014.050
91015.050
91016.050
91017.050
91018.050
91019.050
91020.010
91022.001
91023.050
91030.050
91032.050
91033.050
91034.050
91035.050
91036.050
91037.050
91038.050
91039.050
91040.050
91041.050
91042.050
91043.050
91044.050
91045.050
91046.050
91047.050
91048.050
91049.050
91050.050
91051.050
91052.050
91053.050
91054.050
91055.050
91056.050
91057.050
91058.050
91059.050
91060.050
91061.050
91062.050
91063.050
91064.050
91065.050
91066.050
91067.050
91068.050
91069.050
91070.050
91071.050
VWR® PESTINORM®, Gas Chromatography (GC) Certified Reference Material
Standards
Gaschromatographie-Standards
Due to its excellent separation capacity and its high sensitivity gas chromatography is a widely used technique for analytical purposes. In order to get proper identification and quantification results of the analysed compounds, reference standards are needed. These PESTINORM® reference substances can be used for the characterisation of GC column properties. In addition they may also be used for the identification of unknown compounds in a gas chromatogram or as standards in quantitative GC analysis.
- All products are CRM acc ISO/IEC 17025 and ISO 17034
- All products supplied with a Certificate of Analysis and batch-specific chromatogram
- Assay typically min 99,0% in many cases over 99,5%
- 24 month shelf life
Intended uses:
- For calibration of TLC, GC-FID, GC-TCD, GC-ECD, GC-MS, GC-MS-MS, LC-UV, LC-MS and LC-MS-MS
- Validation of analytical methods
- Preparation of 'working reference samples'
- Detection limit and linearity studies