91021.050EA
205
EUR
InStock
91021.050
91024.050
91025.050
91026.050
91027.050
91028.050
91029.050
91031.050
91029.010
91031.010
91021.010
91024.010
91026.001
91027.010
91028.010
91029.001
91072.050
91073.010
91073.050
91074.010
91074.050
91076.010
91076.050
91078.010
91078.050
91080.010
91080.050
91082.010
91082.050
91083.002
91083.010
91083.050
91087.002
91087.010
91087.050
91088.001
91088.010
91088.050
91089.001
91090.001
91090.010
91090.050
91091.001
91091.002
91091.010
91072.010
92156.001
91257.250
91255.001
91251.010
91254.001
91249.100
91252.100
91250.001
91253.001
VWR® PESTINORM®, Fatty Acid Methyl Esters (FAME) Standards for GC
Standards
Gaschromatographie-Standards
Avantor offers an extensive range of FAME standards as Certified Reference Materials under the well-known PESTINORM® brand.
- All products are CRM acc ISO/IEC 17025 and ISO 17034
- All products are NEAT
- 24 month shelf life
Esterification of fatty acids is an important tool to determine content and characterise fats and oils in food, especially for measurement by GC methods. The simple derivatisation to create the methyl ester analogue is also used widely as a preliminary technique for assessing the quality and purity of biofuels. VWR® offers a comprehensive range of fatty acid methyl esters (FAME) Certified Reference Materials for calibration of GC instruments, for validation of analytical methods, preparation of 'working reference samples' or for detection limit and linearity studies.
Zertifizierungen: ISO 17025 / 17034