VWR® PESTINORM®, GC Multiresidue Pesticide Standard Kit of 203 components grouped in 9 sEPArate mixtures, CRMs
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
91112.001EA 3630 EUR InStock
91112.001
VWR® PESTINORM®, GC Multiresidue Pesticide Standard Kit of 203 components grouped in 9 sEPArate mixtures, CRMs
Standards Organische Standards
A kit for food testing, comprising nine separate mixtures, all Certifified Reference Materials, totalling 203 components. All mixture are dissioved in toluene or toluene-rich solvents
All mixes are supplied with detailed certificate of analysis, chromatogram and 100% shelf-life. The CoAs provide full traceability of preparation, including the dissolution of the raw materials (with their purity), the preparation of the intermediate solutions and the preparation of the final solutions.
Zertifizierungen: Certified Reference Material (CRM) acc. ISO 17025 / ISO 17034
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge