VWR® PESTINORM®, GC Multiresidue Pesticide Standard (Mix 6) - 18 components, CRM
Lieferant: VWR Chemicals
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
91118.001EA 464 EUR InStock
91118.001
VWR® PESTINORM®, GC Multiresidue Pesticide Standard (Mix 6) - 18 components, CRM
Standards Organische Standards
A standard mixture, typically used for food testing, containing the following components: 100 ug/ml each of Acrinathrin [CAS:101007-06-1] ; Anthraquinone [CAS:84-65-1] ; cis-Bifenthrin [CAS:82657-04-3] ; Bioallethrin [CAS:584-79-2] ; Cyfluthrin [CAS:68359-37-5] ; lambda-Cyhalothrin [CAS:91465-08-6] ; Cypermethrin [CAS:52315-07-8] ; Deltamethrin [CAS:52918-63-5] ; Fenvalerate [CAS:51630-58-1] ; Flucythrinate [CAS:70124-77-5] ; tau-Fluvalinate [CAS:102851-06-9] ; cis-Permethrine [CAS:61949-76-6] ; trans-Permethrine [CAS:61949-77-7] ; Phenothrin [CAS:26002-80-2] ; Resmethrin [CAS:10453-86-8] ; Tefluthrin [CAS:79538-32-2] ; Tetramethrin [CAS:7696-12-0] ; Transfluthrin [CAS:118712-89-3] in Toluene
All mixes are supplied with detailed certificate of analysis, chromatogram and 100% shelf-life. The CoAs provide full traceability of preparation, including the dissolution of the raw materials (with their purity), the preparation of the intermediate solutions and the preparation of the final solutions.
Zertifizierungen: Certified Reference Material (CRM) acc. ISO 17025 / ISO 17034
Achtung! Bitte beachten: Storage temperature: Freezer (–20 °C)
Order Now
PRODUKT
TECHNISCHE DATEN
WEITERE INFORMATIONEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge