VWR®, 1H,1H,2H,2H-Perfluoro-1-dodecanol, Certified Reference Material (CRM)
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
91135.001EA 298 EUR InStock
91135.001
VWR®, 1H,1H,2H,2H-Perfluoro-1-dodecanol, Certified Reference Material (CRM)
Standards Organische Standards
Per- and polyfluoroalkyl substances (PFAS) are amongst the primary emerging contaminants of concern. The detection and quantification of known PFAS and the discovery of unknown PFAS substances has never been more important.
VWR® is supplying a comprehensive range of standards and CRMs to enable this environmental analysis.
Zertifizierungen: Certified Reference Material (CRM) ISO 17025 / ISO 17034
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge