VWR®, Multi-element calibration standard-3, 10 Elements: 10 ug/ml each of Sb, Au, Hf, Ir, Pd, Pt, Rh, Ru, Te, Sn in 10% HCl/1% HNO₃
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
91212.180EA 207 EUR InStock
91212.180
VWR®, Multi-element calibration standard-3, 10 Elements: 10 ug/ml each of Sb, Au, Hf, Ir, Pd, Pt, Rh, Ru, Te, Sn in 10% HCl/1% HNO₃
Standards Kationischer Standards Mehrelement-Standards
Zertifizierungen: ISO 17025/17034
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge