VWR®, 6020 Interference Check Solution B for ICP/MS, 11 components in 5% HNO₃
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
91213.180EA 242 EUR InStock
91213.180
VWR®, 6020 Interference Check Solution B for ICP/MS, 11 components in 5% HNO₃
Standards Kationischer Standards Mehrelement-Standards
11 components: Cr 20 ug/ml, Co 20 ug/ml, Cu 20 ug/ml, Mn 20 ug/ml, Ni 20 ug/ml, V 20 ug/ml, As 10 ug/ml, Cd 10 ug/ml, Se 10 ug/ml, Zn 10 ug/ml, Ag 5 ug/ml in 5% HNO₃
Zertifizierungen: ISO 17025/17034
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge