VWR®, Agilent/ Varian Environmental Calibration Standard 25 components in 10% HNO₃
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
91220.180EA 506 EUR InStock
91220.180
VWR®, Agilent/ Varian Environmental Calibration Standard 25 components in 10% HNO₃
Standards Kationischer Standards Mehrelement-Standards
25 components: Ca 1000 ug/ml, Fe 1000 ug/ml, K 1000 ug/ml, Mg 1000 ug/ml, Na 1000 ug/ml, Ag 10 ug/ml, Al 10 ug/ml, As 10 ug/ml, Ba 10 ug/ml, Be 10 ug/ml, Cd 10 ug/ml, Co 10 ug/ml, Cr 10 ug/ml, Cu 10 ug/ml, Mn 10 ug/ml, Mo 10 ug/ml, Ni 10 ug/ml, Pb 10 ug/ml, Sb 10 ug/ml, Se 10 ug/ml, Tl 10 ug/ml, V 10 ug/ml, Zn 10 ug/ml, Th 10 ug/ml, U 10 ug/ml in 10% HNO₃
Zertifizierungen: ISO 17025/17034
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge