VWR®, ICP multi-element standard solution VI 30 components in 5% HNO₃
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
91268.180EA 449 EUR InStock
91268.180
VWR®, ICP multi-element standard solution VI 30 components in 5% HNO₃
Standards Kationischer Standards Mehrelement-Standards
30 components: Ag 10 mg/l, Al 10 mg/l, As 100 mg/l, B 100 mg/l, Ba 10 mg/l, Be 100 mg/l, Bi 10 mg/l, Ca 1000 mg/l, Cd 10 mg/l, Co 10 mg/l, Cr 10 mg/l, Cu 10 mg/l, Fe 100 mg/l, Ga 10 mg/l, K 10 mg/l, Li 10 mg/l, Mg 10 mg/l, Mn 10 mg/l, Mo 10 mg/l, Na 10 mg/l, Ni 10 mg/l, Pb 10 mg/l, Rb 10 mg/l, Se 100 mg/l, Sr 10 mg/l, Te 10 mg/l, Tl 10 mg/l, U 10 mg/l, V 10 mg/l, Zn 100 mg/l in 5% HNO₃
Zertifizierungen: ISO 17025/17034
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge