VWR®, Azoxystrobin, Certified Reference Material
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
91463.0010EA 39 EUR InStock
91463.0010
VWR®, Azoxystrobin, Certified Reference Material
Standards Organische Standards
Pesticides are usually chemical or biological substances that are used to prevent, destroy or mitigate any pests. Pesticides are released into the environemnet in order to eliminate these pests, but residues from these toxic chemicals also end up in the air, water, and even in food.
Our pesticide standards help you to accurately monitor soil, water and food quality in compliance with national and international regulations.
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge