Sigma-Aldrich colour chart, analytical standard, Supelco®
Lieferant: SUPELCO
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
91711-1EA
91711-1EAEA 288 EUR InStock
91711-1EA
Sigma-Aldrich colour chart, analytical standard, Supelco®
Standards Farbreferenzen und Spektroskopie-Standards
Colour reference table for solids.
The Sigma-Aldrich Colour Chart has been developed for internal use in quality control of all the Sigma-Aldrich production sites. Considering our huge portfolio of different chemical products, the common colour system should bring clarity in the jungle of colours and be simple to apply in the daily practice. Furthermore it should be based upon an accepted colour system; communicable, reproducible and applicable everywhere.
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge