Biotin picolyl azide
Lieferant: BIOTIUM
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
92186
92186.EA 509 EUR InStock
92186.
Biotin picolyl azide
Proteinkonjugate und Markierungen
Biotin picolyl azide reacts with alkyne to form 1,2,3-triazole by 1,3-dipolar Huisgen cycloaddition through the use of a much lower copper (I) concentration without sacrificing reaction efficiency.
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge