L-Glutamic acid, certified reference material, TraceCERT®, Supelco®
Lieferant: SUPELCO
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
95436-100MG
95436-100MGEA 86.2 EUR InStock
95436-100MG
L-Glutamic acid, certified reference material, TraceCERT®, Supelco®
Standards Organische Standards
L-Glutamic acid has been used as a reference material in the development of quantitative nuclear magnetic resonance technique to evaluate the purity of organic molecules.
An excitatory amino acid neurotransmitter that is an agonist at all subtypes of glutamate receptors (metabotropic, kainate, NMDA, and AMPA).
Zertifizierungen: This certified reference material (CRM) is produced and certified in accordance with ISO/IEC 17025 and ISO 17034. This CRM is traceable to primary material from an NMI, e.g. NIST or NMIJ. Certified content by quantitative NMR incl. uncertainty and expiry date are given on the certificate.
Verpackung: Bottomless glass bottle. Contents are inside inserted fused cone.
https://qa.digitalassets.avantorsciences.com/adaptivemedia/rendition?id=a76bc25790383dd1260e4be371f5ff1e76ae5c6d&vid=a76bc25790383dd1260e4be371f5ff1e76ae5c6d&prid=web&clid=SAPDAM
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge