Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Ammoniaklösung 27,0 - 30,0% NH3 basis NF, FCC, strong, J.T.Baker®

Lieferant: AVANTOR PERFORMANCE MATERIALS

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

Gefahr

9726-02EA 100.84 EUR InStock
9726-02 9726-05 9726-R
Ammoniaklösung 27,0 - 30,0% NH3 basis NF, FCC, strong, J.T.Baker®
Ammoniaklösung
pH adjusters are used to adjust the pH of formulations. This category includes acids that are notoriously difficult to source as truly cGMP-produced and amino acids that most companies do not fully test for EP, BP and JP. Avantor provides all that and more.


Formel: NH₃
MW: 35.05 g/mol
Siedepunkt: 32…37.7 °C (1013 hPa)
Melting Pt: –60…–57.5 °C
Dichte: 0.88…0.91 g/cm³ (20 °C)
Storage Temperature: Raumtemperatur
MDL Number: MFCD00066650
CAS-Nummer: 1336-21-6
EINECS: 215-647-6
UN: 2672
ADR: 8 III
Order Now
Spezifikation Testergebnisse
Food GMP Manufactured Product Passes Test
Meets N.F. Requirements = 0.02 %
GMP Manufactured Product Passes Test
Meets F.C.C. Requirements 27.0 - 30.0 %
CAUTION: For Manufacturing, processing or repackaging = 5 ppm
Bulk Pharmaceutical Chemical Passes Test
Bulk Food Chemical Passes Test
NF - Identification Passes Test
NF - Nonvolatile Residue ≤0.02 %
NF - Readily Oxidizable Substances Passes Test
NF - Assay (NH₃)(NF) 27.0 - 30.0 %
FCC - Identification Passes Test
FCC - Nonvolatile Residue ≤0.02 %
FCC - Readily Oxidizable Substances Passes Test
FCC - Assay (NH₃)(NF) 27.0 - 30.0 %
FCC - Lead (Pb) (mg/kg) ≤0.5
No Class 1,2,3 or other solvents are used or produced in the
manufacturing or purification of the product.
Elemental Impurities (USP (232), EP 5.20) - Information on elemental
impurities for this product is available on the associated Product
Regulatory Data Sheet and elemental impurity profile report.

 

PRODUKT

TECHNISCHE DATEN

WEITERE INFORMATIONEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge