J.T.Baker®, Cycle-Tap Sampler
Lieferant: Avantor
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
9815-05EA 0 EUR InStock
9815-05
J.T.Baker®, Cycle-Tap Sampler
Probenehmer Probenehmer für Flüssigkeiten
Cycle-Tap sampler is a device for sampling small quantities of solvent.
Designed to eliminate solvent exposure to the atmosphere when used with a Luer lock syringe.
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge