Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

VWR®, Density Standards, Certified Reference Materials, Referenced at 25 °C

Lieferant: VWR Chemicals

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:


99653.600EA 84 EUR InStock
99653.600 99654.600 99655.600 99656.600 99657.600 99660.600 99661.600 99662.600 99663.600 99664.600 99665.600
VWR®, Density Standards, Certified Reference Materials, Referenced at 25 °C
Standards Dichtestandards
VWR® ISO 17025 / ISO 17034 dual certified density standards are for the verification of instruments used to measure density of materials which are fluid at the desired test temperature.

  • ISO 17025 / ISO 17034 certified under UKAS accreditation
  • Density tested in strict accordance with ASTM D1480, a primary method for density determination
  • Density standards range between 0,6654 - 1,6300 g/ml
  • Ensures full compliance to ASTM and IP test method protocols
  • Shelf life between 12 / 24 months


Order Now

 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge