Siliconentschäumer GAMMA EGT SAG 471 zur Produktion
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
997002.6002EA 630 EUR InStock
997002.6002
Siliconentschäumer GAMMA EGT SAG 471 zur Produktion
Siliconentschäumer
Silicone antifoam liquid , sterilised by gamma irradiation. SAG 471 antifoam is for use in both aqueous and non-aqueous systems, imparting outstanding durability in the control of foam over time. Highly effective (even at low concentrations), it offers cost-efficient foam control for a variety of applications, including aerobic fermentation of antibiotics involving actinomycetales organisms, glycol-based coolants, heat exchange systems, adhesives, sealants, vaccines, and others. SAG 471 antifoam is self-dispersing in water.
  • Maximization of fermentor capacities
  • Excellent durability
  • Excellent foam control performance in fermentation processes
  • Self-dispersing in water- suitability under sterilization
Order Now
PRODUKT
TECHNISCHE DATEN
WEITERE INFORMATIONEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge