Multi-component alcohol mixture for calibration, Cerilliant®, Supelco®
Lieferant: SUPELCO
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
A-127-6X1.2ML
A-127-6X1.2MLEA 129 EUR InStock
A-127-6X1.2ML
Multi-component alcohol mixture for calibration, Cerilliant®, Supelco®
Standards Organische Standards
A multicomponent calibration kit containing acetone, ethanol, isopropanol, and methanol at concentrations of 100, 250, 500, 1000, 2000, and 4000 μg/mL. This kit is suitable for calibration of blood alcohol testing methods using GC Headspace.
  • SNAP-N-SPIKE®/SNAP-N-SHOOT®
Achtung! Bitte beachten: Storage temperature: 2 to 8 °C (refrigerator)
https://qa.digitalassets.avantorsciences.com/adaptivemedia/rendition?id=b623cebe99dc8f28540ef8357c0104444f829c45&vid=b623cebe99dc8f28540ef8357c0104444f829c45&prid=web&clid=SAPDAM
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge