A10144.09
A10144.18
A10144.30
A10144.09EA
71.9
EUR
InStock
A10144.09
A10144.18
A10144.30
2,3-Dihydroxybenzoesäure ≥98%
2,3-Dihydroxybenzoic acid
Purity: 98%
CAS No.: 303-38-8
Molecular Formula: C7H6O4
Formula Weight: 154.12
Melting Point: 204-208°
Beilstein Reference: 2209117