A10163.30
A10163.0B
A10163.0I
AB119388.0250
A10163.30EA
43.3
EUR
InStock
A10163.30
A10163.0B
A10163.0I
ABCRAB119388.0250
L(+)-Kaliumnatriumtartrat Tetrahydrat ≥99%
L(+)-Kaliumnatriumtartrat Tetrahydrat
Purity: 99%
CAS No.: 6381-59-5
Molecular Formula: C4H4KNaO6.4H2O
Formula Weight: 282.23 (210.29anhy)
Physical Form: Crystals
Density: 1.77
Appearance: White
Odour: Odourless
pH: 6.0-8.5
Melting Point: 70-80°
Solubility: Soluble in water. Insoluble in alcohol
Beilstein Reference: 6113568
Merck Reference: 14,7670
Optical Rotation: +21° (c=1 in water)
MDL No.: MFCD00150989
Baking powder, medicine (cathartic), component of Fehling's solution, silvering mirrors