A10173.18
A10173.09
A10173.18EA
107
EUR
InStock
A10173.18
A10173.09
Trifluormethansulfonsäure ≥98%
Trifluoromethanesulphonic acid
Purity: 98+%
CAS No.: 1493-13-6
Molecular Formula: CHF3O3S
Formula Weight: 150.07
Flash Point: None
Refractive Index: 1.3270
Odour: Pungent
Melting Point: -40°
Boiling Point: 161-162°
Beilstein Reference: 1812100
Merck Reference: 14,9676
UN No.: 3265
MDL No.: MFCD00007514