A10185.06
A10185.14
A10185.06EA
87
EUR
InStock
A10185.06
A10185.14
3-Amino-6-chlorpyridazin 98%
3-Amino-6-chlorpyridazin
Purity: 98%
CAS No.: 5469-69-2
Molecular Formula: C4H4ClN3
Formula Weight: 129.55
Melting Point: ca 218° dec.
Beilstein Reference: 110918