A10208.09
A10208.18
A10208.30
A10208.09EA
50.9
EUR
InStock
A10208.09
A10208.18
A10208.30
α-Brom-4-fluortoluol ≥97%
α-Brom-4-fluortoluol
Purity: 97%
CAS No.: 459-46-1
Molecular Formula: C7H6BrF
Formula Weight: 189.03
Flash Point: 66°(150°F)
Refractive Index: 1.5470
Odour: Pungent
Boiling Point: 84-85°/15mm
Beilstein Reference: 636507
UN No.: 3265
MDL No.: MFCD00000359