A10228.22
A10228.09
A10228.14
A10228.22EA
107
EUR
InStock
A10228.22
A10228.09
A10228.14
4-Brom-2-fluoranilin ≥98%
4-Brom-2-fluoranilin
Purity: 98+%
CAS No.: 367-24-8
Molecular Formula: C6H5BrFN
Formula Weight: 190.01
Flash Point: 104°(219°F)
Melting Point: 38-42°
Boiling Point: 103-108°/13mm
Beilstein Reference: 2081089
UN No.: 2811
MDL No.: MFCD00010221