A10272.09
A10272.18
A10272.09EA
42
EUR
InStock
A10272.09
A10272.18
2,5-Difluoranilin ≥98%
2,5-Difluoranilin
Purity: 98%
CAS No.: 367-30-6
Molecular Formula: C6H5F2N
Formula Weight: 129.11
Flash Point: 68°(154°F)
Refractive Index: 1.5130
Odour: Characteristic
Melting Point: 13-14°
Boiling Point: 171-174°
Beilstein Reference: 2802549
UN No.: 2941
MDL No.: MFCD00007651