A10275.0B
A10275.14
A10275.30
A10275.0BEA
475
EUR
InStock
A10275.0B
A10275.14
A10275.30
2-Bromethanol ≥97%
2-Bromethanol
Purity: 97%
CAS No.: 540-51-2
Molecular Formula: C2H5BrO
Formula Weight: 124.97
Flash Point: 110°(230°F)
Refractive Index: 1.4930
Melting Point: -80°
Boiling Point: 149-150°
Beilstein Reference: 878140
Merck Reference: 14,3792
UN No.: 2922
MDL No.: MFCD00002827