A10341.06
A10341.14
A10341.06EA
48.3
EUR
InStock
A10341.06
A10341.14
3',5'-Bis(trifluormethyl)acetophenon ≥98%
3',5'-Bis(trifluormethyl)acetophenon
Purity: 98%
CAS No.: 30071-93-3
Molecular Formula: C10H6F6O
Formula Weight: 256.15
Flash Point: 82°(179°F)
Refractive Index: 1.4230
Boiling Point: 85°/8mm
Beilstein Reference: 2384789
MDL No.: MFCD00009910