A10372.0F
A10372.AE
A10372.0FEA
164
EUR
InStock
A10372.0F
A10372.AE
2-Ethylhexylamin 98%
2-Ethylhexylamin
Purity: 98%
CAS No.: 104-75-6
Molecular Formula: C8H19N
Formula Weight: 129.25
Flash Point: 50°(122°F)
Refractive Index: 1.4300
Odour: Amine-like
Melting Point: -76°
Boiling Point: 169°
Beilstein Reference: 1209249
UN No.: 2276
MDL No.: MFCD00008148