A10400.06
A10400.09
A10400.06EA
88.8
EUR
InStock
A10400.06
A10400.09
6-Bromhexannitril ≥97%
6-Bromhexannitril
Purity: 97%
CAS No.: 6621-59-6
Molecular Formula: Br(CH2)5CN
Formula Weight: 176.06
Flash Point: 110°(230°F)
Refractive Index: 1.4775
Boiling Point: 134-135°/15mm
Beilstein Reference: 1744172
UN No.: 3276
MDL No.: MFCD00013834