A10425.14
A10425.22
A10425.36
AB113800.0025
A10425.14EA
53.2
EUR
InStock
A10425.14
A10425.22
A10425.36
ABCRAB113800.0025
Azobenzol ≥97%
Azobenzol
Purity: 97+%
CAS No.: 103-33-3
Molecular Formula: C12H10N2
Formula Weight: 182.23
Melting Point: 65-69°
Boiling Point: 293°
Merck Reference: 14,917
UN No.: 3077
MDL No.: MFCD00003022