A10474.0B
A10474.18
A10474.30
A10474.0BEA
272
EUR
InStock
A10474.0B
A10474.18
A10474.30
L(-)-Menthol ≥99%
L(-)-Menthol
Purity: 99%
CAS No.: 2216-51-5
Molecular Formula: C10H20O
Formula Weight: 156.27
Flash Point: 93°(199°F)
Melting Point: 42-45°
Boiling Point: 212-216°
Beilstein Reference: 1902293
Merck Reference: 14,5837
Optical Rotation: -50° (c=10 in ethanol)
MDL No.: MFCD00062979